Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CX3CL1/Fractalkine Chemokine Domain Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 8 publications
$172.84 - $1184.00
Specifications
Antigen | CX3CL1/Fractalkine |
---|---|
Clone | 81513 |
Dilution | Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB3651SP
|
R&D Systems
MAB3651SP |
25 μg |
Each for $172.84
|
|
|||||
MAB3651100
|
R&D Systems
MAB3651100 |
100 μg |
Each for $532.44
|
|
|||||
MAB3651500
|
R&D Systems
MAB3651500 |
500 μg |
Each for $1,184.00
|
|
|||||
Description
CX3CL1/Fractalkine Monoclonal specifically detects CX3CL1/Fractalkine in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CX3CL1/Fractalkine | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
P78423 | |
6376 | |
NS0 and E. coli-derived recombinant human CX3CL1/Fractalkine Chemokine Domain Gln25-Arg339 (Ser199Asn) Accession # P78423 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human CX3CL1/Fractalkine Chemokine Domain in direct ELISAs and Western blots. |
81513 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ABCD-3, C3Xkine, chemokine (C-X3-C motif) ligand 1, CX3C membrane-anchored chemokine, CXC3, CXC3C, FKN, fractalkine, Neurotactin, neurotactin), NTNsmall inducible cytokine subfamily D (Cys-X3-Cys), member 1 (fractalkine, NTTSmall-inducible cytokine D1, SCYD1C-X3-C motif chemokine 1, small inducible cytokine subfamily D (Cys-X3-Cys), member-1 | |
CX3CL1 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title