Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CX3CL1/Fractalkine Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NBP149539
Description
CX3CL1/Fractalkine Polyclonal specifically detects CX3CL1/Fractalkine in Human, Mouse, Gerbil samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
CX3CL1/Fractalkine | |
Polyclonal | |
Unconjugated | |
PBS and 30% Glycerol with 0.05% Sodium Azide | |
ABCD-3, C3Xkine, chemokine (C-X3-C motif) ligand 1, CX3C membrane-anchored chemokine, CXC3, CXC3C, FKN, fractalkine, Neurotactin, neurotactin), NTNsmall inducible cytokine subfamily D (Cys-X3-Cys), member 1 (fractalkine, NTTSmall-inducible cytokine D1, SCYD1C-X3-C motif chemokine 1, small inducible cytokine subfamily D (Cys-X3-Cys), member-1 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Gerbil | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
1.15 mg/mL | |
Western Blot reported in scientific literature (PMID 31524247), Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:250, Immunohistochemistry-Frozen reported in scientific literature | |
P78423, O35188 | |
CX3CL1 | |
A genomic peptide made to an internal region of the human CX3CL protein (within residues 20-150). [Swiss-Prot P78423] | |
0.1 mL | |
Chemokine Research, Cytokine Research, GPCR, Immunology, Innate Immunity, Virology Bacteria and Parasites | |
6376 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction