Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CX3CL1 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA129026

Catalog No. PA129026


Add to cart

Description

Description

PA1-29026 detects CX3CL1 from mouse samples. PA1-29026 has been successfully used in immunohistochemistry, immunoprecipitation, Western blot applications. The PA1-29026 immunogen is: Recombit protein - mouse CX3CL1

Specifications

Specifications

CX3CL1
Polyclonal
Unconjugated
Cx3cl1
Liquid
CX3CL1
Recombit protein - mouse CX3CL1
Protein A
RUO
Antibody
Polyclonal
Mouse
Immunohistochemistry, Immunoprecipitation, Western Blot
1 mg/mL
PBS with 0.1% sodium azide; pH 7.2
O35188
AB030188, ABCD-3, AI848747, CX3C, Cxc3, D8Bwg0439e, Scyd1
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
20312
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit