Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CX3CL1, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA130943

Catalog No. PA130943

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA1-30943 detects CX3CL1 from human, mouse samples.PA1-30943 has been successfully used in immunohistochemistry (paraffin), Western blot applications.The PA1-30943 immunogen is: Purified recombinant human fractalkine, chemokine domain, expressed in E. coli.Avoid freeze/thaw cycles
Specifications

Specifications

CX3CL1
Polyclonal
Unconjugated
CX3CL1
Purified
CX3CL1
Purified recombinant human fractalkine, chemokine domain, expressed in E. coli.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine
Immunohistochemistry (Paraffin), Western Blot
0.1mg/mL
PBS with 5% trehalose and no preservative
P78423, O35188
ABCD-3, C3Xkine, CXC3, CXC3C, NTN, NTT, SCYD1, fractalkine, neurotactin
Goat
IgG
50μg
Store at -20°C or Store at -80°C if preferred
Primary
6376
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit