Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CX3CR1, PerCP, Clone: 528728, R&D Systems™

Manufacturer:  R&D Systems FAB5204C025

 View more versions of this product

Catalog No. FAB5204C025


Add to cart

Description

Description

Host Species: Mouse
Clone: 528728
Isotype: IgG1
Species Reactivity: Hu
Immunogen: E. coli-derived recombinant human CX3CR1. Met1-Thr31, Leu91-Lys103, Thr168-Thr195, Lys257-Leu273

Applications:

Flow Cytometry

Specifications

Specifications

CX3CR1
528728
Flow Cytometry 10 uL/10^6 cells
NP_001328
Beta chemokine receptor-like 1, CCRL1, chemokine (C-C) receptor-like 1, chemokine (C-X3-C motif) receptor 1, chemokine (C-X3-C) receptor 1, CMKbRL1, CMK-BRL1, CMK-BRL-1, CMKDR1, CX3C chemokine receptor 1, Fractalkine receptor, G protein-coupled receptor 13, GPR13G-protein coupled receptor 13, GPRV28, V28, V28C-X3-C CKR-1
Mouse
IgG1
25 Tests
Primary
1524
Human
Flow Cytometry
PerCP
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
CX3CR1
E. coli-derived recombinant human CX3CR1, Met1-Thr31, Leu91-Lys103, Thr168-Thr195, Lys257-Leu273, Accession # NP_001328
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects human CX3CR1 in direct ELISAs.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit