Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CXCL10/IP-10/CRG-2, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP145664

Catalog No. NBP145664

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

CXCL10/IP-10/CRG-2
0.5 mg/mL
Western Blot
P48973
C7, chemokine (C-X-C motif) ligand 10, crg-2, gIP-10, IFI10, INP10, IP-10, mob-1, SCYB10
Rabbit
IgG
0.1 mg
Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity, Signal Transduction
Primary
3627
Western Blot
Unconjugated
PBS (pH 7.2), 1.0% BSA and 50% Glycerol with 0.02% Thimerosal
Affinity Purified
CXCL10
E. coli-expressed recombinant rat CXCL10/INP10
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only