Learn More
anti-CXCL10/IP-10/CRG-2, Polyclonal, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP177875
Catalog No. NBP177875
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
CXCL10/IP-10/CRG-2
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Purified
CXCL10
This protein-A purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a recombinant protein raised in yeast, corresponding to the 83 amino acids of the mature bovine CVCL10/INP10 protein.
Protein A purified
RUO
Store at 4C. Do not freeze.
Polyclonal
This antibody is specific for bovine CVCL10/INP10 protein. A BLAST analysis was used to suggest cross-reactivity with CVCL10/INP10 from bovine, goat, and sheep sources based on 100% identity with immunogen.
ELISA, Western Blot
Western Blot 1 ug/ml, ELISA 1:500
NP_001040016
C7, chemokine (C-X-C motif) ligand 10, crg-2, gIP-10, IFI10, INP10, IP-10, mob-1, SCYB10
Rabbit
IgG
0.1 mg
Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity, Signal Transduction
Primary
3627
Bovine, Goat, Sheep