Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CXCL12/SDF-1 beta, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP145849

Catalog No. NBP145849

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

CXCL12/SDF-1 beta
0.5 mg/mL
Western Blot
P48061
hSDF-1beta, SDF 1beta, SDF1B
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
6387
Western Blot
Unconjugated
PBS (pH 7.2), 0.5% BSA and 30% Glycerol with 0.01% Thimerosal
Affinity Purified
CXCL12
Synthetic peptide surrounding amino acid 74 of human SDF1 beta.
11 kDa
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only