Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL13, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA127553

Catalog No. PIPA127553

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Chemotactic for B-lymphocytes but not for T-lymphocytes. monocytes and neutrophils. Does not induce calcium release in B-lymphocytes. Binds to BLR1/CXCR5. B lymphocyte chemoattractant. independently cloned and named Angie. is a CXC chemokine strongly expr

PA1-27553 detects CXCL13 from human samples.PA1-27553 may be successfully used in Western blot applications.The immunogen is a synthetic peptide corresponding to human Small inducible cytokine B13.
Specifications

Specifications

CXCL13
Polyclonal
PBS with no preservative
O43927
ANGIE, ANGIE2, BCA-1, BCA1, BLC, BLR1L, SCYB13
Chicken
IgY
50μg
Store at -20°C
Primary
10563
Western Blot
Unconjugated
CXCL13
Purified
CXCL13
Synthetic peptide corresponding to human Small inducible cytokine B13.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit