Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL16 Rat anti-Mouse, Clone: 142417, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.

Rat Monoclonal Antibody

$114.50 - $839.00

Specifications

Antigen CXCL16
Immunogen E. coli-derived recombinant mouse CXCL16 Asn27-Pro114 Accession # Q8BSU2
Clone 142417
Applications Western Blot, Neutralization, ELISA
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB503SP
View Documents
R&D Systems
MAB503SP
25 ug
Each for $114.50
Add to cart
 
MAB503100
View Documents
R&D Systems
MAB503100
100 ug
Each for $377.00
Add to cart
 
MAB503500 Available on GSA/VA Contract for Federal Government customers only.
View Documents
R&D Systems
MAB503500
500 ug
Each for $839.00
Add to cart
 
Description

Description

CXCL16 Monoclonal antibody MAB503 Specifically detects CXCL16 in Mouse samples. It is validated for ELISA Capture, Neutralization, Western Blotting
Specifications

Specifications

CXCL16
142417
Unconjugated
Q8BSU2
Monoclonal
Primary
Detects mouse CXCL16 in ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human CXCL1, 2, 3, 5, 6, 7, 8, 9, 10, 11, 12, 13, 14, recombinant mouse CXCL1, 2, 6, 9, 10, 11, 12, 13, 14, 15, recombinant rat CXCL1, 2, 3, 5, or recombinant porcine CXCL8 is observed.
Protein A or G purified from hybridoma culture supernatant
E. coli-derived recombinant mouse CXCL16 Asn27-Pro114 Accession # Q8BSU2
Western Blot, Neutralization, ELISA
Purified
Rat
IgG2a
58191
RUO
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit