Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL9 Monoclonal Antibody (49106), PE, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA523628
Description
In ELISAs, does not cross-react with recombinant mouse (rm) CXCL9 or recombinant human CXCL10.
CXCL9 Monoclonal antibody specifically detects CXCL9 in Human samples. It is validated for Flow Cytometry.Specifications
CXCL9 | |
Monoclonal | |
PE | |
CXCL9 | |
BB139920; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 9; CMK; crg-10; CXC; CXC motif chemokine; C-X-C motif chemokine 9; C-X-C motif chemokine ligand 9; CXCL; CXCL9; gamma interferon-induced monokine; gamma-interferon-induced monokine; HuMIG; M119; Mig; monokine induced by gamma interferon; monokine induced by interferon gamma; monokine induced by interferon-gamma; MuMIG; protein m119; SCYB9; small inducible cytokine B subfamily (Cys-X-Cys), member 9; small inducible cytokine B9; small-inducible cytokine B9 | |
Mouse | |
Protein A/G | |
RUO | |
4283 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
49106 | |
Buffered saline solution with BSA and 0.09% sodium azide | |
Q07325 | |
CXCL9 | |
E. coli-derived recombinant human CXCL9/MIG Thr23-Thr125 | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction