Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CXCR1/IL-8 RA, PE, Clone: 42705, R&D Systems™

Manufacturer:  R&D Systems FAB330P025

 View more versions of this product

Catalog No. FAB330P025


Add to cart

Description

Description

Host Species: Mouse
Clone: 42705
Isotype: IgG2A
Species Reactivity: Hu
Immunogen: NS0 mouse myeloma cell line transfected with human CXCR1/IL-8 RA. Met1-Leu350

Applications:

Flow Cytometry

Specifications

Specifications

CXCR1/IL-8 RA
42705
Flow Cytometry 10 uL/10^6 cells
AAA59159
CD128, CD181, CD181 antigen, CDw128aC-C, chemokine (C-X-C motif) receptor 1, CKR-1, CMKAR1, C-X-C chemokine receptor type 1, CXC-R1, CXCR-1, High affinity interleukin-8 receptor A, IL-8 receptor type 1, IL-8R A, IL8R1, IL8RA, IL-8RA, IL8RAC-C-CKR-1, IL8RBA, interleukin 8 receptor, alpha, interleukin-8 receptor type 1, interleukin-8 receptor type A
Mouse
IgG2a
25 Tests
Primary
3577
Human
Flow Cytometry
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
CXCR1
NS0 mouse myeloma cell line transfected with human CXCR1/IL-8 RA, Met1-Leu350, Accession # AAA59159
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects human CXCR1/IL-8 RA transfectants but not the parental cell line. It does not cross-react with human CXCR2.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit