Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CXCR3, PE, Clone: 220803, R&D Systems™

Manufacturer:  R&D Systems FAB1685P025

 View more versions of this product

Catalog No. FAB1685P025


Add to cart

Description

Description

Host Species: Rat
Clone: 220803
Isotype: IgG2A
Species Reactivity: Ms
Immunogen: CHO Chinese hamster ovary cell line transfected with mouse CCR3. Met1-Leu367

Applications:

Flow Cytometry

Specifications

Specifications

CXCR3
220803
Flow Cytometry 10 uL/10^6 cells
AAC40163
CD183, CD183 antigen, chemokine (C-X-C motif) receptor 3, chemokine (C-X-C) receptor 3, CKR-L2IP-10 receptor, CMKAR3, C-X-C chemokine receptor type 3, CXCR3, CXC-R3, CXCR-3, G protein-coupled receptor 9CD182, GPR9, GPR9Mig-R, Interferon-inducible protein 10 receptor, IP10 receptor, IP10-R, Mig receptor, MigR
Rat
IgG2a
25 Tests
Primary
2833
Mouse
Flow Cytometry
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
CXCR3
CHO Chinese hamster ovary cell line transfected with mouse CXCR3, Met1-Leu367, Accession # AAC40163
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects mouse CXCR3.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit