Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CXCR3, PerCP, Clone: 49801, R&D Systems™

Manufacturer:  R&D Systems FAB160C025

 View more versions of this product

Catalog No. FAB160C025


Add to cart

Description

Description

Host Species: Mouse
Clone: 49801
Isotype: IgG1
Species Reactivity: Hu
Immunogen: NS0 mouse myeloma cell line transfected with human CXCR3. Met1-Leu368

Applications:

Flow Cytometry

Specifications

Specifications

CXCR3
49801
Flow Cytometry 10 uL/10^6 cells
P49682
CD183, CD183 antigen, chemokine (C-X-C motif) receptor 3, chemokine (C-X-C) receptor 3, CKR-L2IP-10 receptor, CMKAR3, C-X-C chemokine receptor type 3, CXCR3, CXC-R3, CXCR-3, G protein-coupled receptor 9CD182, GPR9, GPR9Mig-R, Interferon-inducible protein 10 receptor, IP10 receptor, IP10-R, Mig receptor, MigR
Mouse
IgG1
25 Tests
Primary
2833
Human
Flow Cytometry
PerCP
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
CXCR3
NS0 mouse myeloma cell line transfected with human CXCR3, Met1-Leu368, Accession # P49682
Protein A or G purified from ascites
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects human CXCR3.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit