Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CXCR4, Alexa Fluor 700, Clone: 12G5, R&D Systems™

Manufacturer:  R&D Systems FAB170N100

 View more versions of this product

Catalog No. FAB170N100


Add to cart

Description

Description

Host Species: Mouse
Clone: 12G5
Isotype: IgG2A
Species Reactivity: Hu
Immunogen: CP-MAC-infected SUP-T1 human T cell lymphoblastic lymphoma line.

Applications:

Flow Cytometry

Specifications

Specifications

CXCR4
12G5
Flow Cytometry 5 uL/10^6 cells
P61073
CD184, chemokine (C-X-C motif) receptor 4, chemokine (C-X-C motif), receptor 4 (fusin), C-X-C chemokine receptor type 4, CXC-R4, CXCR-4, D2S201ESDF-1 receptor, FB22, Fusin, HM89NPYRL, HSY3RR, LAP3, LCR1, LESTRCD184 antigen, Leukocyte-derived seven transmembrane domain receptor, leukocyte-derived seven-transmembrane-domain receptor, lipopolysaccharide-associated protein 3, neuropeptide Y receptor Y3, NPY3RWHIM, NPYR, NPYY3R, seven transmembrane helix receptor, seven-transmembrane-segment receptor, spleen, Stromal cell-derived factor 1 receptor
Mouse
IgG2a
100 Tests
Primary
7852
Human
Flow Cytometry
Alexa Fluor 700
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
CXCR4
CP-MAC-infected SUP-T1 human T cell lymphoblastic lymphoma line, Accession # P61073
Protein A or G purified from ascites
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects human CXCR4.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit