Learn More
CXCR4 (M02), Mouse anti-Human, Clone: 1F8, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CXCR4.
Supplier: Abnova Corporation H00007852M02
Description
This gene encodes a CXC chemokine receptor specific for stromal cell-derived factor-1. The protein has 7 transmembrane regions and is located on the cell surface. It acts with the CD4 protein to support HIV entry into cells and is also highly expressed in breast cancer cells. Mutations in this gene have been associated with WHIM (warts, hypogammaglobulinemia, infections, and myelokathexis) syndrome. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq
Sequence: MEGISIYTSDNYTEEMGSGDYDSMKEPCFREENANFNKIFLPTIYSSpecifications
CXCR4, | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC020968 | |
CXCR4 | |
CXCR4 (AAH20968, 1 a.a. ∼ 46 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Western Blot | |
1F8 | |
Mouse monoclonal antibody raised against a partial recombinant CXCR4. | |
CXCR4 | |
CD184/D2S201E/FB22/HM89/HSY3RR/LAP3/LCR1/LESTR/NPY3R/NPYR/NPYRL/NPYY3R/WHIM | |
Mouse | |
Affinity chromatography | |
RUO | |
7852 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.