Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCR4, Mouse, Clone: 2H5, Abnova™

Mouse monoclonal antibody raised against a partial recombinant CXCR4.

Manufacturer:  Abnova Corporation H00007852M01

Catalog No. 89-002-724


Add to cart

Description

Description

This gene encodes a CXC chemokine receptor specific for stromal cell-derived factor-1. The protein has 7 transmembrane regions and is located on the cell surface. It acts with the CD4 protein to support HIV entry into cells and is also highly expressed in breast cancer cells. Mutations in this gene have been associated with WHIM (warts, hypogammaglobulinemia, infections, and myelokathexis) syndrome. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq

Sequence: MEGISIYTSDNYTEEMGSGDYDSMKEPCFREENANFNKIFLPTIYS
Specifications

Specifications

CXCR4,
2H5
Mouse monoclonal antibody raised against a partial recombinant CXCR4.
CXCR4
Liquid
CXCR4
CXCR4 (AAH20968, 1 a.a. ~ 46 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 ug
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA, Western Blot
Unconjugated
In 1X PBS, pH 7.4
BC020968
CD184|D2S201E|FB22|HM89|HSY3RR|LAP3|LCR1|LESTR|NPY3R|NPYR|NPYRL|NPYY3R|WHIM
Mouse
IgG2a Kappa
RUO
Primary
7852
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit