missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCR4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 22 publications
$204.00 - $482.50
Specifications
Antigen | CXCR4 |
---|---|
Dilution | Western Blot 1:5000, Flow Cytometry, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:200-1:1000, Immunohistochemistry-Frozen |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10074396T
|
Novus Biologicals
NB10074396SS |
0.025 mL |
Each for $204.00
|
|
|||||
NB10074396
|
Novus Biologicals
NB10074396 |
0.1 mL |
Each for $482.50
|
|
|||||
Description
CXCR4 Polyclonal specifically detects CXCR4 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
CXCR4 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
Rabbit | |
Adaptive Immunity, Cancer, Chemokines and Cytokines, Cytokine Research, GPCR, Growth and Development, Immunology, Innate Immunity, Metastasis, Neuronal Cell Markers, Signal Transduction, Stem Cells | |
Whole antisera with 0.05% Sodium Azide | |
CD184, chemokine (C-X-C motif) receptor 4, chemokine (C-X-C motif), receptor 4 (fusin), C-X-C chemokine receptor type 4, CXC-R4, CXCR-4, D2S201ESDF-1 receptor, FB22, Fusin, HM89NPYRL, HSY3RR, LAP3, LCR1, LESTRCD184 antigen, Leukocyte-derived seven transmembrane domain receptor, leukocyte-derived seven-transmembrane-domain receptor, lipopolysaccharide-associated protein 3, neuropeptide Y receptor Y3, NPY3RWHIM, NPYR, NPYY3R, pCXCR4, seven transmembrane helix receptor, seven-transmembrane-segment receptor, spleen, Stromal cell-derived factor 1 receptor | |
CXCR4 | |
IgG | |
Unpurified |
Western Blot 1:5000, Flow Cytometry, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:200-1:1000, Immunohistochemistry-Frozen | |
Polyclonal | |
Antisera | |
RUO | |
Human, Mouse, Rat | |
P70658 | |
7852 | |
A synthetic peptide made to a C-terminal region of the human CXCR4 protein (within residues 300-352). [Swiss-Prot P61073] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'