Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CXCR5, APC, Clone: 51505, R&D Systems™

Manufacturer:  R&D Systems FAB190A100

 View more versions of this product

Catalog No. FAB190A100


Add to cart

Description

Description

Host Species: Mouse
Clone: 51505
Isotype: IgG2B
Species Reactivity: Hu
Immunogen: NS0 mouse myeloma cell line transfected with human CXCR5. Met1-Phe372

Applications:

Flow Cytometry

Specifications

Specifications

CXCR5
51505
Flow Cytometry 10 uL/10^6 cells
P32302
BLR1, BLR1MDR-15, Burkitt lymphoma receptor 1, Burkitt lymphoma receptor 1, GTP binding protein (chemokine (C-X-C motif)receptor 5), Burkitt lymphoma receptor 1, GTP-binding protein, CD185, CD185 antigen, chemokine (C-X-C motif) receptor 5, C-X-C chemokine receptor type 5, CXCR5, CXC-R5, MDR15CXCR-5, MGC117347, Monocyte-derived receptor 15
Mouse
IgG2b
100 Tests
Primary
643
Human
Flow Cytometry
APC
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
CXCR5
NS0 mouse myeloma cell line transfected with human CXCR5, Met1-Phe372, Accession # P32302
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Stains human CXCR5 transfectants but not the parental cell lines in flow cytometry. Does not cross-react with human CXCR2, CXCR3, or CXCR4 transfectants.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit