Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CXCR5, Fluorescein, Clone: 614641, R&D Systems™

Manufacturer:  R&D Systems FAB6198F025

 View more versions of this product

Catalog No. FAB6198F025


Add to cart

Description

Description

Host Species: Rat
Clone: 614641
Isotype: IgG2A
Species Reactivity: Ms
Immunogen: E. coli-derived recombinant mouse CXCR5. Met1-Pro57

Applications:

Flow Cytometry

Specifications

Specifications

CXCR5
614641
Flow Cytometry 10 uL/10^6 cells
Q04683
BLR1, BLR1MDR-15, Burkitt lymphoma receptor 1, Burkitt lymphoma receptor 1, GTP binding protein (chemokine (C-X-C motif)receptor 5), Burkitt lymphoma receptor 1, GTP-binding protein, CD185, CD185 antigen, chemokine (C-X-C motif) receptor 5, C-X-C chemokine receptor type 5, CXCR5, CXC-R5, MDR15CXCR-5, MGC117347, Monocyte-derived receptor 15
Rat
IgG2a
25 Tests
Primary
643
Mouse
Flow Cytometry
Fluorescein
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
CXCR5
E. coli-derived recombinant mouse CXCR5, Met1-Pro57, Accession # Q04683
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects recombinant mouse (rm) CXCR5 in direct ELISAs. In this format, no cross-reactivity with rmCXCR7 is observed.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit