Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CXCR7/RDC-1, PerCP, Clone: 358426, R&D Systems™

Manufacturer:  R&D Systems FAB42271C100

 View more versions of this product

Catalog No. FAB42271C10


Add to cart

Description

Description

Host Species: Mouse
Clone: 358426
Isotype: IgG2A
Species Reactivity: Hu
Immunogen: NS0 mouse myeloma cell line transfected with human CXCR7/RDC?1. Met1-Lys362 (Gly131Ser)

Applications:

Flow Cytometry

Specifications

Specifications

CXCR7/RDC-1
358426
Flow Cytometry 10 uL/10^6 cells
AAA62370
ACKR3, chemokine (C-X-C motif) receptor 7, Chemokine orphan receptor 1CMKOR1C-X-C chemokine receptor type 7, CMKOR1, CXCR7, CXC-R7, CXCR-7, G protein-coupled receptor, GPR159, GPR159G-protein coupled receptor 159, RDC-1, RDC1G-protein coupled receptor RDC1 homolog
Mouse
IgG2a
100 Tests
Primary
57007
Human
Flow Cytometry
PerCP
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
CXCR7
NS0 mouse myeloma cell line transfected with human CXCR7/RDC-1, Met1-Lys362 (Gly131Ser), Accession # AAA62370
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects human CXCR7/RDC-1 in direct ELISAs. In flow cytometry, reacts specifically with five distinct human CXCR7 transfectants, but does not react with their respective parental lines or mouse CXCR7 transfectants. In flow cytometry, also reacts with monocytes expressing CXCR7, but does not react with MCF-7 cells which have been reported to have surface-expressing CXCR7 using clone 11G8. Due to the conflicting reports published, use of clone 358426 may result in an underestimation of CXCR7 expression on certain cell types.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit