Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CXXC1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP191816

Catalog No. NBP191816


Add to cart

Description

Description

CXXC1 Polyclonal antibody specifically detects CXXC1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

CXXC1
Unconjugated
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
CFP1CXXC-type zinc finger protein 1, CGBPPHD finger and CXXC domain-containing protein 1, CpG binding protein, cpG-binding protein, CXXC finger 1 (PHD domain), CXXC finger protein 1, DNA-binding protein with PHD finger and CXXC domain, hCGBP, HsT2645, PCCX12410002I16Rik, PHF185830420C16Rik, SPP1, ZCGPC1, zinc finger, CpG binding-type containing 1
Rabbit
IgG
0.1 ml
Neuroscience
Primary
30827
Human
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:10-1:20
Affinity Purified
CXXC1
This antibody was developed against Recombinant Protein corresponding to amino acids:VKVKHVKRREKKSEKKKEERYKRHRQKQKHKDKWKHPERADAKDPASLPQCLGPGCVRPAQPSSKYCSDDCGMKLAANRIYEILPQRIQQWQQSPCIAEEHGKKLLERIRREQQSARTRLQEMERRFHELEAIILRAKQQ
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only