Learn More
CXXC4 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA526391
Catalog No. PIPA526391
Add to cart
Description
Description
This antibody is predicted to react with rat based on sequence homology.
CXXC4 acts as a negative regulator of the Wnt signaling pathway via its interaction with DVL1.
Specifications
Specifications
CXXC4
Polyclonal
Unconjugated
CXXC4
Liquid
CXXC4
KLH conjugated synthetic peptide between 158-185 amino acids from the C-terminal region of human CXXC4
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q9H2H0, Q6NXI8
IDAX, CXXC finger 4, CXXC-type zinc finger protein 4, Dvl-binding protein IDAX (inhibition of the Dvl and Axin complex), inhibition of the Dvl and axin complex protein
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
319478, 80319