Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome C1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525257

Catalog No. PIPA525257

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse based on sequence homology.

This is the heme-containing component of the cytochrome b-c1 complex, which accepts electrons from Rieske protein and transfers electrons to cytochrome c in the mitochondrial respiratory chain.
Specifications

Specifications

Cytochrome C1
Polyclonal
PBS with 0.09% sodium azide
P08574
UQCR4, complex III subunit 4, complex III subunit IV, cytochrome b-c1 complex subunit 4, cytochrome c1, heme protein, mitochondrial, ubiquinol-cytochrome-c reductase complex cytochrome c1 subunit
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1537
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
CYC1
Liquid
CYC1
KLH conjugated synthetic peptide between 259-287 amino acids from the C-terminal region of human CYC1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit