Learn More
Cyclin A Rabbit anti-Human, Mouse, Rat, Sheep, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 06138
Catalog No. 06-138-MI
Add to cart
Description
Description
Detect Cyclin A with Anti-Cyclin A Antibody (Rabbit Polyclonal Antibody), that has been shown to work in immunoprecipitation, western blot, immunocytochemistry.
Specifications
Specifications
Cyclin A
Unconjugated
Purified
Rabbit
IgG
200μg
Primary
NM_001237
Immunocytochemistry, Immunoprecipitation, Western Blot
Lyophilized
CCNA2; Cyclin-A2; CCN1; Cyclin-A; CCNA
peptide corresponding amino acid residues 417-432 (C-KHGVSLLNPPETL-N-L)
Protein G chromatography
−20°C for 24 months
Polyclonal
Human, Murine, Rat, Sheep