Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclin B1 Mouse anti-Human, Mouse, Clone: GNS3 (8A5D12), MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05373
Description
Anti-Cyclin B1 Antibody, clone GNS3 (8A5D12) is a Mouse Monoclonal Antibody for detection of Cyclin B1 also known as G2/mitotic-specific cyclin B1 and has been validated in immunoprecipitation and western blotting.Specifications
Cyclin B1 | |
Monoclonal | |
Unconjugated | |
NM_031966 | |
Mouse | |
Protein G chromatography | |
Primary | |
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG Mix |
Immunoprecipitation, Western Blot | |
GNS3 (8A5D12) | |
Protein G purified mouse IgGs in storage buffer containing 0.1MTris-glycine, pH 7.4, 0.15MNaCl, 0.05% sodium azide. Frozen at −20°C. | |
CCNB1, CCNB | |
Recombinant human cyclin B1 | |
200 μg | |
Human, Mouse | |
Purified |
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'
For Research Use Only