Learn More
Cyclin D Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 06137
Catalog No. 06-137-MI
Add to cart
Description
Description
Anti-Cyclin D Antibody is a high quality Rabbit Polyclonal Antibody for the detection of Cyclin D and has been validated in immunocytochemistry, immunoprecipitation and western blotting.
Specifications
Specifications
Cyclin D
Unconjugated
Purified
Rabbit
IgG
200μg
Primary
NM_053056
Immunocytochemistry, Immunoprecipitation, Western Blot
Protein A Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide as a preservative.
CCND1; D11S287E; U21B31; PRAD1; BCL1
peptide corresponding the 11 C-terminal amino acids. residues 285-295, of human Cyclin D1 (CTPTDVRDVDI)
Protein A purfied
−20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles.
Polyclonal
Human, Murine, Rat