Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclin D1/2 Mouse anti-Human, Hamster, Mouse, Rat, Clone: 5D4, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05362
Description
Anti-Cyclin D1/2 Antibody, clone 5D4 is a high quality Mouse Monoclonal Antibody for the detection of Cyclin D1/2 and has been validated in immunocytochemistry, immunoprecipitation and western blotting.Specifications
Cyclin D1/2 | |
Monoclonal | |
Unconjugated | |
NM_053056, NM_001759 | |
Mouse | |
Protein A purfied | |
Primary | |
−20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles. | |
IgG2a |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
5D4 | |
Protein A Purified immunoglobulin in Protein G Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105MNaCl, 0.035% sodium azide as a preservative. | |
CCND1, D11S287E, U21B31, PRAD1, BCL1 | |
Recombinant human cyclin D1 | |
100 μg | |
Hamster, Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only