Learn More
MilliporeSigma™ anti-Cyclin D1, Clone: ,
Manufacturer:  MilliporeSigma™ 05815
Catalog No. 05-815-MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Cyclin D1
Unconjugated
NM_053056
CCND1, D11S287E, U21B31, PRAD1, BCL1
GST fusion protein cleaved with Factor Xa and gel purified, corresponding to complete mouse Cyclin D1
Protein G purfied
−20°C for up to 24 months.
Monoclonal
Immunocytochemistry, Immunoprecipitation, Western Blot
0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Purified
Murine
IgG1
100μg
Primary
Murine, Rat