Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin D3 Monoclonal Antibody, Clone: DCS2.2, Invitrogen™

Cyclin D3 Antibody (DCS2.2), MA1-12298, from Invitrogen™. Species Reactivity: Human, Mouse, Rat; Applications: Immunohistochemistry, Immunoprecipitation, Western Blot.

Manufacturer:  Invitrogen MA112298

Catalog No. 01-670-991

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Cyclin D3 Antibody (DCS2.2), MA1-12298, from Invitrogen. Species Reactivity: Human, Mouse, Rat; Applications: Immunohistochemistry, Immunoprecipitation, Western Blot.

The immunogen for the anti-h-Cyclin D3 was a recombinant full length Cyclin D3 protein recognizing a 34 kD protein. Cyclin D3 is a G1 cyclin closedly related to Cyclins D1 and D2 phase progression. Like Cyclin D1 and Cyclin D2, it is activated by the same mechanisms and can phosphorylate pRB when associated with cdk4 and cdk6.
Host Species: Mouse
Cross Reactivity: Human, Mouse, Rat
Isotype: IgG1
Clone: DCS2.2
Immunogen: Full length recombinant Cyclin D3 protien.
Formulation: 100µg purified antibody in PBS and 0.08% sodium azide
Synonyms: RP5-973N23.3
Uniprot Accession: P30281, P30282, P48961
Entrez GeneID: 12445, 25193, 896
Gene Symbol: CCND3
This product detects Cyclin D3 in Human, Mouse and Rat samples. This product has been successfully used in Immunohistochemistry, Immunoprecipitation and Western Blot procedures. The immunogen is Full length recombinant Cyclin D3 protien..
References:
Test: IHC, Dilution: 1:100, Species: Hu, Reference: Cyclin D1-negative mantle cell lymphoma: a clinicopathologic study based on gene expression profiling. (2005 Dec 15) Blood, Vol: 106, 13, 4315-21, Fu K, Weisenburger DD, Greiner TC, Dave S, Wright G, Rosenwald A, Chiorazzi M, Iqbal J, Gesk S, Siebert R, De Jong D, Jaffe ES, Wilson WH, Delabie J, Ott G, Dave BJ, Sanger WG, Smith LM, Rimsza L, Braziel RM, Müller-Hermelink HK, Campo E, Gascoyne RD, Staudt LM, Chan WC; Pubmed ID: 16123218
Specifications

Specifications

Cyclin D3
Monoclonal
Unconjugated
P30281, P30282, P48961
RP5-973N23.3
Murine
IgG1
RUO
Primary
Monoclonal
Human, Murine, Rat
Immunohistochemistry, Immunoprecipitation, Western Blot
DCS2.2
100μg purified antibody in PBS and 0.08% sodium azide
Purified
CCND3
Full length recombinant Cyclin D3 protien.
100μg
Store at -20°C, Avoid Freeze/Thaw Cycles
Primary
12445, 25193, 896
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit