Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Cyclin M3, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232133

Catalog No. NBP232133


Add to cart

Description

Description

Cyclin M3 Polyclonal antibody specifically detects Cyclin M3 in Human samples. It is validated for Immunoprecipitation.
Specifications

Specifications

Cyclin M3
1.0 mg/mL
Immunoprecipitation 2 - 10 mcg/mg lysate
NP_060093.3
ACDP3cyclin-M3, ancient conserved domain protein 3, Ancient conserved domain-containing protein 3, cyclin M3, Cyclin-M3, DKFZp434I1016, FLJ20018, metal transporter CNNM3
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human
Immunoprecipitation
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
CNNM3
The immunogen this antibody was made to, maps to a region between residue 575 to 625 of human Cyclin M3 using the numbering given in entry NP_060093.3 (GeneID 26505).
Immunogen affinity purified
RUO
Primary
26505
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only