Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin O Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA520262

Catalog No. PIPA520262


Add to cart

Description

Description

This antibody will recognize both isoforms of Cyclin-O. A suggested positive control is mouse bladder tissue lysate. PA5-20262 can be used with blocking peptide PEP-0377.

Specifications

Specifications

Cyclin O
Polyclonal
Unconjugated
CCNO
Liquid
CCNO
A 14 amino acid synthetic peptide from near the amino terminus of human Cyclin O
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
P22674, P0C242
CCNO
Rabbit
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
10309, 218630, 499528
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit