Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclin T1 Sheep anti-Human, Mouse, Polyclonal, MilliporeSigma™
Sheep Polyclonal Antibody
Supplier: MilliporeSigma 061289
Description
Use Anti-Cyclin T1 Antibody (Sheep Polyclonal Antibody) validated in western blotting to detect Cyclin T1 also known as CDK9-associated C-type protein, cyclin C-related protein.Specifications
Cyclin T1 | |
Polyclonal | |
Sheep polyclonal serum with 0.05% sodium azide. | |
Sheep | |
Unpurified | |
Primary | |
Human, Mouse | |
Serum |
Western Blot | |
Unconjugated | |
CCNT1; CCNT; CYCT1; CycT1; Cyclin-T; HIVE1 | |
His-tagged recombinant protein corresponding to human Cyclin T1. | |
100 μL | |
NP_001231.2 | |
−20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction