Learn More
Cyclin T1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA131486
Catalog No. PA131486
Add to cart
Description
Description
PA1-31486 detects Cyclin T1 from mouse, human, rat samples. PA1-31486 has been successfully used in immunohistochemistry (paraffin tissue), immunoprecipitation, ELISA, Western blot applications. The PA1-31486 immunogen is: Cyclin T1 peptide corresponding to an internal region of the human protein conjugated to KLH. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
The protein encoded by this gene belongs to the highly conserved cyclin family, whose members are characterized by a dramatic periodicity in protein abundance through the cell cycle. Cyclins function as regulators of CDK kinases. Different cyclins exhibit distinct expression and degradation patterns which contribute to the temporal coordination of each mitotic event. This cyclin tightly associates with CDK9 kinase, and was found to be a major subunit of the transcription elongation factor p-TEFb. The kinase complex containing this cyclin and the elongation factor can interact with, and act as a cofactor of human immunodeficiency virus type 1 (HIV-1) Tat protein, and was shown to be both necessary and sufficient for full activation of viral transcription. This cyclin and its kinase partner were also found to be involved in the phosphorylation and regulation of the carboxy-terminal domain (CTD) of the largest RNA polymerase II subunit.
Specifications
Specifications
Cyclin T1
Polyclonal
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
O60563, Q9QWV9
CCNT, CYCT1, HIVE1
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Mouse, Rat
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
CCNT1
Liquid
CCNT1
Cyclin T1 peptide corresponding to an internal region of the human protein conjugated to KLH.
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
12455, 315291, 904