Learn More
CYP27B1 Rabbit anti-Human, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA526065
Catalog No. PIPA526065
Add to cart
Description
Description
CYP27B1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. The protein encoded by this gene localizes to the inner mitochondrial membrane where it hydroxylates 25-hydroxyvitamin D3 at the 1alpha position. This reaction synthesizes 1alpha,25-dihydroxyvitamin D3, the active form of vitamin D3, which binds to the vitamin D receptor and regulates calcium metabolism. Thus this enzyme regulates the level of biologically active vitamin D and plays an important role in calcium homeostasis. Mutations in this gene can result in vitamin D-dependent rickets type I.
Specifications
Specifications
CYP27B1
Polyclonal
Unconjugated
CYP27B1
Liquid
CYP27B1
KLH conjugated synthetic peptide between 482-508 amino acids from the C-terminal region of human CYP27B1
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Rat
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
O15528, O35084, O35132
CP2B, CYP1, CYP1alpha, CYP27B, P450c1, PDDR, VDD1, VDDR, VDDRI, VDR, 1alpha(OH)ase, 25 hydroxyvitamin D3-1-alpha hydroxylase, 25-OHD-1 alpha-hydroxylase, 25-hydroxyvitamin D-1 alpha hydroxylase, mitochondrial, VD3 1A hydroxylase, calcidiol 1-monooxygenase, cytochrome P450 subfamily XXVIIB polypeptide 1, cytochrome P450C1 alpha, cy
Rabbit
IgG
200μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
114700, 13115, 1594