Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP2C8 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA515215

Catalog No. PIPA515215

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CYP2C8 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and its expression is induced by phenobarbital. The enzyme is known to metabolize many xenobiotics, including the anticonvulsive drug mephenytoin, benzo(a)pyrene, 7-ethyoxycoumarin, and the anti-cancer drug taxol.
Specifications

Specifications

CYP2C8
Polyclonal
PBS with 0.09% sodium azide
P10632
S-mephenytoin 4-hydroxylase; P450 IIC2; P450 MP-12/MP-20; P450 form 1; CYPIIC8; Cytochrome P450 2C8; CYP2C8
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1558
Western Blot
Unconjugated
CYP2C8
Liquid
CYP2C8
KLH conjugated synthetic peptide between 74-105 amino acids from the N-terminal region of human CYP2C8
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit