Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP2R1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA515061

Catalog No. PIPA515061

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CYP2R1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This enzyme is a microsomal vitamin D hydroxylase that converts vitamin D into the active ligand for the vitamin D receptor. A mutation in CYP2R1 gene has been associated with selective 25-hydroxyvitamin D deficiency.
Specifications

Specifications

CYP2R1
Polyclonal
PBS with 0.09% sodium azide
Q6VVX0
Vitamin D 25-hydroxylase; Cytochrome P450 2R1; CYP2R1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
120227
Immunohistochemistry, Western Blot
Unconjugated
CYP2R1
Liquid
CYP2R1
KLH conjugated synthetic peptide between 464-496 amino acids from the C-terminal region of human CYP2R1
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit