Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP3A7 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527976

Catalog No. PIPA527976


Add to cart

Description

Description

PA5-27976 targets CYP3A7 in WB applications and shows reactivity with Human samples. The PA5-27976 immunogen is recombit fragment corresponding to a region within amino acids 222 and 489 of Human CYP3A7. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene, CYP3A7, encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This enzyme hydroxylates testosterone and dehydroepiandrosterone 3-sulphate, which is involved in the formation of estriol during pregcy. The enzyme also metabolizes some drugs such as aflatoxin B1. This gene is part of a cluster of cytochrome P450 genes on chromosome 7q21.1. Transcript variants have been described, but it is not known whether these transcripts are normally produced.
Specifications

Specifications

CYP3A7
Polyclonal
Unconjugated
CYP3A7
Liquid
CYP3A7
Recombit fragment corresponding to a region within amino acids 222 and 489 of Human CYP3A7
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.8 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
P24462
CP37, CYPIIIA7, P450-HFLA, aryl hydrocarbon hydroxylase, cytochrome P450 3A7, cytochrome P450, subfamily IIIA, polypeptide 7, cytochrome P450-HFLA, flavoprotein-linked monooxygenase, microsomal monooxygenase, xenobiotic monooxygenase
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
1551
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit