Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP4X1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA515051

Catalog No. PIPA515051

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CYP4X1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. The expression pattern of a similar rat protein suggests that this protein may be involved in neurovascular function in the brain.
Specifications

Specifications

CYP4X1
Polyclonal
Unconjugated
CYP4X1
Liquid
CYP4X1
KLH conjugated synthetic peptide between 157-183 amino acids from the N-terminal region of human CYP4X1
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
2.0 mg/mL
PBS with 0.09% sodium azide
Q8N118
CYPIVX1; Cytochrome P450 4X1; CYP4X1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
260293
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit