Learn More
CYR61 Rabbit anti-Human, Rabbit, DyLight® 488, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA522764
Catalog No. PIPA522764
Add to cart
Description
Description
This antibody is predicted to react with rat and mouse based on 88% sequence homology.
CYR61 is a cysteine-rich, heparin-binding, growth factor belonging to the CCN family of secreted regulatory proteins. It stimulates chondrogensis and angiogenesis, playing an important role in cartilage and bone formation, as well as tumor growth and vascularization.
Specifications
Specifications
CYR61
Polyclonal
DyLight™ 488
CYR61
Liquid
CYR61
A synthetic peptide made to the human CYR61 protein sequence (between residues 150-250).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Rabbit
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
sodium borate with no preservative
O00622
CCN1, GIG1, IGFBP10, CCN family member 1, IBP-10, IGF-binding protein 10, IGFBP-10, cysteine-rich heparin-binding protein 61, cysteine-rich, anigogenic inducer, 61, insulin-like growth factor-binding protein 10, protein CYR61
Rabbit
IgG
100 μL
4° C, store in dark
Primary
100339657, 3491