Learn More
CYP3A4 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA184227
Catalog No. PIPA184227
Add to cart
Description
Description
This antibody is a specific and potent inhibitor of CYP3A4 activity. Rabbit anti Human cytochrome p450 3A4 antibody recognizes human cytochrome p450 3A4 (CYP3A4).
P450 enzymes are divided into two groups: steroidogenic and Xenobiotic. The latter group is comprised of three families 1 ,2 and 3. The xenobiotic p450 ft.s are involved in most oxidative drug metabolism. Work in this area is ongoing but studies suggest that all known drug metabolism is mediated by family members: CYP3a, CYP2D6, CYP2C9/10, CYP2C19, CYP2E1.
Specifications
Specifications
CYP3A4
Polyclonal
whole serum with no preservative
P08684
CP33, CP34, CYP3A, CYP3A3, HLP, MGC126680, NF-25, P450C3, P450PCN1
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Functional Assay, Western Blot
Unconjugated
CYP3A4
Liquid
CYP3A4
Purified human CYP3A4 expressed in E.Coli.
50 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1576