Learn More
Cytoglobin Mouse anti-Human, Clone: 3A5-2D2, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA522919
Catalog No. PIMA522919
Add to cart
Description
Description
Peptide Sequence: MEKVPGEMEI ERRERSEELS EAERKAVQAM WARLYASCED VGVAILVRFF VNFPSAKQYF SQFKHMEDPL EMERSPQLRK HACRVMGALN TVVENLHDPD KVSSVLALVG KAHALKHKVE PVYFKILSGV ILEVVAEEFA SDFPPETQRA WAKLRGLIYS HVTAAYKEVG WVQQVPNATT PPATLPSSGP
Cytoglobin is a ubiquitously expressed hexacoordinate hemoglobin that may facilitate diffusion of oxygen through tissues, scavenge nitric oxide or other reactive oxygen species, or serve a protective function during oxidative stress (Trent and Hargrove, 2002 [PubMed 11893755]).
Specifications
Specifications
Cytoglobin
Monoclonal
0.5 mg/mL
PBS with no preservative; pH 7.4
Q8WWM9
HGB, STAP, histoglobin, stellate cell activation-associated protein
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
114757
ELISA, Immunocytochemistry, Immunofluorescence
3A5-2D2
Unconjugated
CYGB
Liquid
CYGB
CYGB (AAH29798, 1 a.a. ∽ 190 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human