Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Cytohesin 4, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP219974
Description
Cytohesin 4 Polyclonal specifically detects Cytohesin 4 in Human samples. It is validated for Western Blot.Specifications
Cytohesin 4 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
CYT4PH, SEC7 and coiled-coil domain-containing protein 4, cytohesin 4, cytohesin-4, pleckstrin homology, Sec7 and coiled/coil domains 4, pleckstrin homology, Sec7 and coiled-coil domains 4, PSCD4DJ63G5.1 | |
Rabbit | |
46 kDa | |
0.1 mL | |
Primary | |
Human | |
IgG |
Western Blot | |
0.25mg/mL | |
Western Blot 1:500-1:3000 | |
Q9UIA0 | |
CYTH4 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human PSCD4. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
27128 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only