Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 13 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522065

Catalog No. PIPA522065


Add to cart

Description

Description

PA5-22065 targets Cytokeratin 13 in ICC, IF, IHC (P), and WB applications and shows reactivity with Human samples. The PA5-22065 immunogen is recombit protein fragment contain a sequence corresponding to a region within amino acids 198 and 438 of Cytokeratin 13. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

Cytokeratin 13
Polyclonal
Unconjugated
KRT13
Liquid
KRT13
Recombit protein fragment contain a sequence corresponding to a region within amino acids 198 and 438 of Cytokeratin 13
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.59 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P13646
CK13, K13, CK-13, cytokeratin 13, cytokeratin-13, keratin, type I cytoskeletal 13, keratin-13
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
3860
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit