Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 14, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA128209

Catalog No. PIPA128209

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. They range in molecular weight between ~40kDa and ~

PA1-28209 detects Cytokeratin 14 from human samples.PA1-28209 has been successfully used in immunohistochemistry (paraffin tissue) applications.The PA1-28209 immunogen is: Synthetic peptide (unfortunately, the amino acid sequence is considered to be commercially sensitive) C terminal (Human).
Specifications

Specifications

Cytokeratin 14
Polyclonal
PBS with 1% BSA and 0.1% sodium azide; pH 7.4
P02533
Keratin-14, CK-14, K14 Synonyms Cytokeratin-14 CK-14 Keratin-14 K14, Keratin-14, CK-14
Rabbit
IgG
7mL
Store at 4°C
Primary
3861
Immunohistochemistry (Paraffin)
Unconjugated
KRT14
Purified
KRT14
Synthetic peptide corresponding to the Human C terminal region.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit