Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 18 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528279

Catalog No. PIPA528279


Add to cart

Description

Description

PA5-28279 targets Cytokeratin 18 in IF, IHC (P), and WB applications and shows reactivity with Human samples. The PA5-28279 immunogen is recombit fragment corresponding to a region within amino acids 73 and 300 of Human Cytokeratin 18. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Cytokeratin 18 (Keratin 18, KRT18, type I cytoskeletal 18) belongs to the type A (acidic) subfamily of low molecular weight keratins, exists in combination with cytokeratin 8. Tissues from gastrointestinal tract, respiratory tract and urogenital tract, as well as endocrine and exocrine tissues and mesothelial cells are positive for cytokeratin 18. Further, cytokeratin 18 is found primarily in non-squamous epithelia and is present in a majority of adenocarcinomas and ductal carcinomas, but not in squamous cell carcinomas. Hepatocellular carcinomas express only cytokeratins 8 and 18. Mutations in the cytokeratin 18 gene have been linked to cryptogenic cirrhosis. Two transcript variants encoding the same protein have been found for the cytokeratin 18 gene. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial maligcies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays.
Specifications

Specifications

Cytokeratin 18
Polyclonal
Unconjugated
KRT18
Liquid
KRT18
Recombit fragment corresponding to a region within amino acids 73 and 300 of Human Cytokeratin 18
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.54 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P05783
CYK18, K18, CK-18, cell proliferation-inducing gene 46 protein, cell proliferation-inducing protein 46, cytokeratin 18, cytokeratin-18, keratin, type I cytoskeletal 18, keratin-18
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
3875
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit