Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 19 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
$204.00 - $482.50
Specifications
Antigen | Cytokeratin 19 |
---|---|
Concentration | 1.0 mg/ml |
Dilution | Western Blot 1:1000, Simple Western 1:1000, Flow Cytometry 2-5 ug/million cells, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin 1:500 |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100687SS
|
Novus Biologicals
NB100687SS |
0.025 mL |
Each for $204.00
|
|
|||||
NB100687
|
Novus Biologicals
NB100687 |
0.1 mL |
Each for $482.50
|
|
|||||
Description
Cytokeratin 19 Polyclonal specifically detects Cytokeratin 19 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Cytokeratin 19 | |
Western Blot 1:1000, Simple Western 1:1000, Flow Cytometry 2-5 ug/million cells, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin 1:500 | |
Polyclonal | |
Rabbit | |
Cancer, Cell Biology, Cytoskeleton Markers, Stem Cell Markers | |
PBS and 30% Glycerol with 0.05% Sodium Azide | |
CK19, CK-19, cytokeratin 19, Cytokeratin-19,40-kDa keratin intermediate filament, K19cytokeratin-19, K1CS, keratin 19, keratin, type I cytoskeletal 19, keratin, type I, 40-kd, keratin-19, MGC15366 | |
KRT19 | |
IgG | |
Affinity Purified | |
Human, mouse, and monkey. Rat reactivity reported in scientific literature (PMID: 25401473). |
1.0 mg/ml | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
P08727 | |
3880 | |
Synthetic peptide derived from the C-terminal region of human Cytokeratin 19 (between residues 350-400) [UniProt P08727] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title