Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 19, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA128210

Catalog No. PIPA128210

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Cytokeratins, a group comprising at least 29 different proteins, are characteristic of epithelial and trichocytic cells. Cytokeratin 5, 6 and 8 are members of the type II neutral-to-basic subfamily. Monoclonal antibodies to cytokeratins are specific marke

PA1-28210 detects Cytokeratin 19 from human samples.PA1-28210 has been successfully used in immunohistochemistry (paraffin tissue) applications.The PA1-28210 immunogen is: Synthetic peptide (unfortunately, the amino acid sequence is considered to be commercially sensitive) C terminal (Human).
Specifications

Specifications

Cytokeratin 19
Polyclonal
PBS with 1% BSA and 0.1% sodium azide; pH 7.4
P08727
CK-19, Keratin-19, K19
Rabbit
IgG
7mL
4° C, do not freeze
Primary
3880
Immunohistochemistry (Paraffin)
Unconjugated
KRT19
Liquid
KRT19
Synthetic peptide corresponding to the Human C terminal region.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit