anti-Cytokeratin , PE, Clone: CAM 5.2, BD

Anti-Cytokeratin (CAM 5.2) reagent has a primary reactivity with human keratin proteins that correspond to Moll's peptides No.7 and No.8, Mr 48 and 52kDa, respectively.Cytokeratin 7 and 8 are present on secretory epithelia of normal human tissue but not on stratified squamous epithelium. Anti-Cytokeratin (CAM 5.2) stains most epithelial-derived tissue, including liver, renal tubular epithelium, and hepatocellular and renal cell carcinomas. Anti-Cytokeratin (CAM 5.2) may not react with some squamous cell carcinomas.

$276.00

Specifications

Antigen Cytokeratin
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Host Species Mouse
Clone CAM 5.2
View More Specs
  • Products 1
  • Description & Specifications
Products
Catalog Number Mfr. No. Quantity Conjugate Price Quantity & Availability    

BDB347204

BD
347204
50 Tests PE Each for $276.00
Check Availability
Add to Cart
Descriptions & Specifications

Related Products