Learn More
Cytomegalovirus IE Early Antigen Mouse anti-Virus, Clone: F88G, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA17024
Catalog No. MA17024
Add to cart
Description
Description
MA1-7024 detects the Cytomegalovirus. This antibody is specific for an immediate early antigen present early in the infectious cycle. MA1-7024 has been successfully used in immunofluorescence procedures. The MA1-7024 immunogen is Cytomegalovirus lysate.
Cytomegalovirus (CMV) is an opportunistic pathogen infecting lung, kidney, gut and other organs in situations where an individual is immunologically immature, such as the foetus and neonate. Infection also occurs in immunosuppressed patients e.g. transplant patients, patients undergoing chemotherapy and those with HIV infection. Cytomegalovirus is an enveloped double stranded DNA virus which in humans (HCMV) is commonly known as human herpesvirus 5 (HHV-5).
Specifications
Specifications
Cytomegalovirus IE Early Antigen
Monoclonal
0.1 mg/mL
PBS with 0.1% sodium azide; pH 7.2
CMV
Cytomegalovirus lysate.
Protein A
RUO
Antibody
Monoclonal
Immunofluorescence
F88G
Unconjugated
Liquid
Mouse
IgG1
1 mL
4° C
Primary
Virus